4-Bromopicolinic acid 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Bromopyridine-2-carboxylic acid
OR5568-5G
OR5568-10G
OR5568-25G
76592-664EA
20.32
USD
76592-664
76592-666
76592-668
4-Bromopicolinic acid 97%
4-Bromopicolinic acid
Formula:
C₆H₄BrNO₂ Melting Pt: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426665 CAS Number: 30766-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...