L-Threitol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: L-1,2,3,4-Butanetetrol

OR55453-1G OR55453-250MG
76592-514EA 158 USD
76592-514 76592-512
L-Threitol 98%
L-Threitol
Formula: C₄H₁₀O₄
MW: 122.12 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00064294
CAS Number: 2319-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR