Cyclopentyl methyl ether (CPME) 99% stabilized

Supplier: APOLLO SCIENTIFIC
Danger

OR55202-1L OR55202-2.5L OR55202-500ML
76625-340EA 89.02 USD
76625-340 76625-342 76625-344
Cyclopentyl methyl ether (CPME) 99% stabilized
Cyclopentyl methyl ether (CPME)
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 106 °C
Melting Pt: –134 °C
Density: 0.86 g/cm³ (25 °C)
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD08276401
CAS Number: 5614-37-9
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR