Octadecylphosphonic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: n-Octadecylphosphonic acid, ODPA

OR55172-25G OR55172-5G
76592-316EA 24.46 USD
76592-316 76592-314
Octadecylphosphonic acid 97%
Octadecylphosphonic acid
Formula: C₁₈H₃₉O₃P
Melting Pt: 92…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00015838
CAS Number: 4724-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR