Olive oil, pure, virgin
Supplier: Thermo Scientific Chemicals
41654-0025
41654-0250
41654-5000
TS41654-0025EA
218
USD
TS41654-0025
TS41654-0250
TS41654-5000
Olive oil, pure, virgin
Olive oil
Density:
0.9135 g/cm³ Flash Pt: 225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00131764 CAS Number: 8001-25-0 EINECS: 232-277-0 |
Specification Test Results
Appearance | Clear yellow to yellow-green Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4670 to 1.4710 (20 °C, 589 nm) |
Water | ≤0.1 % |
Absorbance | ≤0.2 (270nm, 1g/100ml cyclohexane) |
Composition | 2.5 to 21.0 % (Linoleic acid) |
Acid value | ≤2.0 mg KOH/g |
Peroxide value | ≤20.0 meq O2/kg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...