Olive oil, pure, refined

Supplier: Thermo Scientific Chemicals

41653-0025 41653-0250 41653-5000
TS41653-0025EA 187 USD
TS41653-0025 TS41653-0250 TS41653-5000
Olive oil, pure, refined
Olive oil
Density: 0.9135 g/cm³
Flash Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00131764
CAS Number: 8001-25-0
EINECS: 232-277-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow-green Oily liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4710 (20 °C, 589 nm)
Water ≤0.1 % (Coulometric)
Absorbance ≤1.20 (1g/100ml cyclohexane) measured at the max. between 264nm and 276nm
Composition ≤0.5 % (delta 7-Stigmasterol)
Acid value ≤0.3 mg KOH/g
Peroxide value ≤10.0 meq O2/kg
Unsaponifiable matter ≤1.5 %
Sesame oil Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR