Hydroxylamine-O-sulfonic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Amino hydrogen sulfate, Amino hydrogen sulphate

32594-0010 32594-0250 32594-1000
TS32594-0010EA 2001 USD
TS32594-0010 TS32594-0250 TS32594-1000
Hydroxylamine-O-sulfonic acid 97%
Hydroxylamine-O-sulphonic acid
Formula: H₃NO₄S
MW: 113.09 g/mol
Melting Pt: 210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011604
CAS Number: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR