(S)-(+)-β-Aminophenethyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol
30757-0010
30757-0050
30757-0250
TS30757-0010EA
34
USD
TS30757-0010
TS30757-0050
TS30757-0250
(S)-(+)-β-Aminophenethyl alcohol 98%
(S)-(+)-β-Aminophenethyl alcohol
Formula:
C₈H₁₁NO Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064404 CAS Number: 20989-17-7 UN: 3263 ADR: 8,III |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 75 to 79 °C |
HPLC | ≥97.5 % |
Specific optical rotation | +24.5° to +26.5° (20 °C, 589 nm) (c=6, CH3OH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...