Theophyllin, anhydrous 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline

25031-0010 25031-0050 25031-2500
TS25031-0010EA 256 USD
TS25031-0010 TS25031-0050 TS25031-2500
Theophyllin, anhydrous 99+%
Theophyllin
Formula: C₇H₈N₄O₂
MW: 180.16 g/mol
Melting Pt: 270…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00079619
CAS Number: 58-55-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 270 to 274 °C
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR