N,N,N',N'-Tetramethylbenzidine 97.5%

Supplier: Thermo Scientific Chemicals

Synonyms: N,n,n,n-tetramethylbenzidine, 4,4'-Bis(dimethylamino)biphenyl

13838-0050 13838-0250
TS13838-0050EA 91 USD
TS13838-0050 TS13838-0250
N,N,N',N'-Tetramethylbenzidine 97.5%
N,N,N',N'-Tetramethylbenzidine
Formula: C₁₆H₂₀N₂
Melting Pt: 192…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00008310
CAS Number: 366-29-0
EINECS: 206-676-5

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
Titration with HClO4 ≥96.5 %
HPLC ≥97.5 %
Specific optical rotation +76° to +83° (20 °C, 589 nm) (c=1, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR