4-(Dimethylamino)benzonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

11578-0050 11578-0250
TS11578-0050EA 40 USD
TS11578-0050 TS11578-0250
4-(Dimethylamino)benzonitrile 95%
4-(Dimethylamino)benzonitrile
Formula: C₉H₁₀N₂
Melting Pt: 70…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00001815
CAS Number: 1197-19-9
EINECS: 214-819-8

Order Now

Specification Test Results

Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 76 °C
Titration with HClO4 ≥94 %
Stabilizer About 500 ppm Nitrocresol + Nitrophenol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR