Hydroxylammonium sulfate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(hydroxylammonium) sulphate, Hydroxylamine sulfate (2:1), Hydroxylamine sulphate, Hydroxylamine sulphate (2:1), Bis(hydroxylammonium) sulfate, Hydroxylamine sulfate
19853-0010
19853-0051
19853-1000
TS19853-0010EA
126
USD
TS19853-0010
TS19853-0051
TS19853-1000
Hydroxylammonium sulfate 99%
Hydroxylammonium sulphate
Formula:
H₈N₂O₆S MW: 164.14 g/mol Melting Pt: 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044869 CAS Number: 10039-54-0 EINECS: 233-118-8 UN: 2865 ADR: 8,III Merck Index: 13,04853 |
Specification Test Results
Appearance | White Crystals |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...