(R)-(+)-Limonene 96% unstabilized

Supplier: Thermo Scientific Chemicals
Danger

17939-0025 17939-0050 17939-5000
TS17939-0025EA 328 USD
TS17939-0025 TS17939-0050 TS17939-5000
(R)-(+)-Limonene 96% unstabilized
(R)-(+)-Limonene
Formula: C₁₀H₁₆
Boiling Pt: 175.5 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.8405 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3,III
Merck Index: 13,05515

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4740 (20 °C, 589 nm)
GC ≥95 %
Relative density (D20/4°C) 0.840 to 0.844
Specific optical rotation +102° to +110° (20 °C, 589 nm) (C=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR