Esculin hydrate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Aesculin, 6,7-Dihydroxycoumarin 6-glucoside hydrate, Esculin

11783-0500 11783-0050 11783-0100
TS11783-0500EA 243 USD
TS11783-0500 TS11783-0050 TS11783-0100
Esculin hydrate 97%
Esculin hydrate
Formula: C₁₅H₁₆O₉·xH₂O
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00149492
CAS Number: 531-75-9
EINECS: 208-517-5

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Specific optical rotation -84° to -87° (20°C, 589 nm) (c=1.5, dioxane/H2O 1:1) (on dry substance)
Titration with NaOH ≥96.0 % (On dry substance)
Bromate (BrO3) ≤0.001 %
Iodate (IO3) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.02 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR