Cyclopentylamine 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aminocyclopentane

11159-0050 11159-0250 11159-1000 11159-5000
TS11159-0050EA 20 USD
TS11159-0050 TS11159-0250 TS11159-1000 TS11159-5000
Cyclopentylamine 99+%
Cyclopentylamine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 108
Density: 0.86 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00001380
CAS Number: 1003-03-8
EINECS: 213-697-3
UN: 3286
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4482 to 1.4502 (20°C, 589 nm)
Water ≤0.3 %
GC ≥99.0 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR