4-Aminosalicylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-2-hydroxybenzoic acid

10462-0050 10462-1000 10462-5000
TS10462-0050EA 20 USD
TS10462-0050 TS10462-1000 TS10462-5000
4-Aminosalicylic acid 99%
4-Aminosalicylic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347
Melting Pt: 150 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007789
CAS Number: 65-49-6
EINECS: 200-613-5
Merck Index: 13,00476

Order Now

Specification Test Results

Appearance Beige Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR