(±)-3-Amino-1,2-propanediol 98%

Supplier: Thermo Scientific Chemicals
Danger

10442-0050 10442-0250 10442-1000 10442-2500
TS10442-0050EA 33 USD
TS10442-0050 TS10442-0250 TS10442-1000 TS10442-2500
(±)-3-Amino-1,2-propanediol 98%
(±)-3-Amino-1,2-propanediol
Formula: C₃H₉NO₂
Boiling Pt: 264…265 °C (739 mmHg)
Density: 1.17 g/cm³
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00008140
CAS Number: 616-30-8
EINECS: 210-475-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4940 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR