3-Methylanthranil acid 98%

Supplier: Thermo Scientific Chemicals
Warning

10397-0100 10397-0500
TS10397-0100EA 88 USD
TS10397-0100 TS10397-0500
3-Methylanthranil acid 98%
3-Methylanthranil acid
Formula: C₈H₉NO₂
Melting Pt: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00007745
CAS Number: 4389-45-1
EINECS: 224-505-2

Order Now

Specification Test Results

Appearance Pink to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR