2-Phenylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminobiphenyl, Biphenyl-2-ylamine, 2-Biphenylylamine

10322-0050 10322-0250
TS10322-0050EA 26 USD
TS10322-0050 TS10322-0250
2-Phenylaniline 98%
2-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 48…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007701
CAS Number: 90-41-5
EINECS: 201-990-9

Order Now

Specification Test Results

Appearance Brown to purple Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR