(-)-Scopolamine n-butylbromide

Supplier: Thermo Scientific Chemicals
Warning

46484-0010 46484-0050
TS46484-0010EA 61 USD
TS46484-0010 TS46484-0050
(-)-Scopolamine n-butylbromide
(-)-Scopolamine n-butylbromide

  • Bioactive

Formula: C₂₁H₃₀BrNO₄
MW: 440.38 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00078561
CAS Number: 149-64-4

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 138 to 142 °C
Loss on drying ≤3 % (105 °C)
Solubility (5% in water) Clear colorless solution
Titration Argentimetric 97.5 to 102.5 %
Specific optical rotation -16° to -22° (20 °C, 589 nm) (5% in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR