Pseudopelletierine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9-Methyl-9-azabicyclo[3.3.1]nonan-3-one

46398-0010 463980050 463980250
TS46398-0010EA 61 USD
TS46398-0010 TS463980050 TS463980250
Pseudopelletierine
Pseudopelletierine

  • Bioactive

Formula: C₉H₁₅NO
MW: 153.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078161
CAS Number: 552-70-5

Order Now

Specification Test Results

Appearance White to yellow to brown Powder
Infrared spectrum Conforms
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR