Sulfasalazine 97%

Supplier: Thermo Scientific Chemicals
Danger

46124-0250 46124-1000 46124-0050
TS46124-0250EA 123 USD
TS46124-0250 TS46124-1000 TS46124-0050
Sulfasalazine 97%
Sulfasalazine
Formula: C₁₈H₁₄N₄O₅S
Melting Pt: 260…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00057363
CAS Number: 599-79-1

Order Now

Specification Test Results

Appearance Light yellow to brownish-yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105°C, 2 h)
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR