Bergapten 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Methoxypsoralen, 4-Methoxyfuro[3,2-g]benzopyran-7-one, 4-Methoxy-7H-furo[3,2-g]chromen-7-one

46066-0010 46066-0050 46066-0250
TS46066-0010EA 59 USD
TS46066-0010 TS46066-0050 TS46066-0250
Bergapten 98%
Bergapten
Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Melting Pt: 190…193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010272
CAS Number: 484-20-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 189 to 193 °C
Loss on drying ≤2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR