Daidzin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

46042-0010 46042-1000
TS46042-0010EA 612 USD
TS46042-0010 TS46042-1000
Daidzin 98%
Daidzin
Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017466
CAS Number: 552-66-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR