Artesunate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Oxo-4-(((3R,5aS,6R,8aS,9R,10R,12R,12aR)-3,6,9-trimethyldecahydro-3H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10-yl)oxy)butanoic acid

46034-0050 46034-0250
TS46034-0050EA 42 USD
TS46034-0050 TS46034-0250
Artesunate
Artesunate
Formula: C₁₉H₂₈O₈
MW: 384.43 g/mol
Melting Pt: 131…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866204
CAS Number: 88495-63-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR