Disodium succinate, anhydrous for biochemistry
Supplier: Thermo Scientific Chemicals
|
Warning
|
45963-1000
45963-5000
TS45963-1000EA
31
USD
TS45963-1000
TS45963-5000
Disodium succinate, anhydrous for biochemistry
Disodium succinate
Formula:
C₄H₄Na₂O₄ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002790 CAS Number: 150-90-3 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.3 % |
UV | at 280 nm A: ≤0.1 |
pH | 6 to 9 (0.4 mol sol) |
Titration with HClO4 | 98.5 to 102 % |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...