(-)-Cytisine 98%

Supplier: Thermo Scientific Chemicals
Danger

45885-0010 45885-2500
TS45885-0010EA 403 USD
TS45885-0010 TS45885-2500
(-)-Cytisine 98%
(-)-Cytisine
Formula: C₁₁H₁₄N₂O
Boiling Pt: 218 °C (3hPa)
Melting Pt: 154…156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00136048
CAS Number: 485-35-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 152 to 158 °C
Loss on drying ≤5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR