Simvastatin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate

45884-0010 45884-0050 45884-0250
TS45884-0010EA 60 USD
TS45884-0010 TS45884-0050 TS45884-0250
Simvastatin 98%
Simvastatin
Formula: C25H38O5
Melting Pt: 127…132 °C
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072007
CAS Number: 79902-63-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 135 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR