Amorolfine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Amorolfin, Ro 14-4767/002, Amorolfine Hydrochloride, Curanail, Loceryl, (2R,6S)-rel-2,6-Dimethyl-4-(2-methyl-3-(4-(tert-pentyl)phenyl)propyl)morpholine hydrochloride, (2R,6S)-rel-4-[3-[4-(1,1-Dimethylpropyl)phenyl]-2-methylpropyl]-2,6-dimethyl-morpholine hydrochloride

45875-0010 45875-0050 45875-2500
TS45875-0010EA 191 USD
TS45875-0010 TS45875-0050 TS45875-2500
Amorolfine hydrochloride 98%
Amorolfine hydrochloride
Formula: C₂₁H₃₅NO
MW: 353.98 g/mol
Melting Pt: 221…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00903738
CAS Number: 78613-38-4
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder or powder
Infrared spectrum Conforms
Melting point 206 to 211 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR