Arbutin 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Hydroxyphenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol, Hydroquinone-beta-D-glucoside
45564-0050
45564-0250
45564-1000
TS45564-0050EA
82
USD
TS45564-0050
TS45564-0250
TS45564-1000
Arbutin 98%
Arbutin
Formula:
C₁₂H₁₆O₇ MW: 272.25 g/mol Melting Pt: 195…201 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016915 CAS Number: 497-76-7 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 196 to 201 °C |
Loss on drying | ≤2 % |
HPLC | ≥97.5 % |
Specific optical rotation | -63° to -67° (20 °C, 589 nm) (c=3, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...