Prednisolone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Prednisolonum

44947-0010 44947-0050 44947-0250
TS44947-0010EA 40 USD
TS44947-0010 TS44947-0050 TS44947-0250
Prednisolone 99%
Prednisolone
Formula: C₂₁H₂₈O₅
MW: 360.45 g/mol
Boiling Pt: 571
Melting Pt: 246 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003649
CAS Number: 50-24-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥98.5 %
Specific optical rotation +113° to +119° (20°C, 589 nm) (c=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR