Diethanolamine ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

44906-0010 44906-2500
TS44906-0010EA 213 USD
TS44906-0010 TS44906-2500
Diethanolamine ACS
Diethanolamine
Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 268.8
Melting Pt: 28
Density: 1.0955 g/cm³
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
UN: 3259
ADR: 8 III
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now

Specification Test Results

Appearance Clear viscous liquid
Water ≤0.15 %
Sulfated ash ≤0.005 %
GC ≤1.0 % (Triethanolamine)
Color scale ≤15 APHA
Apparent equivalent weight 104.0 to 106.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR