Potassium hydrogen carbonate 99.5% for biochemistry

Supplier: Thermo Scientific Chemicals

44801-0025 44801-5000
TS44801-0025EA 231 USD
TS44801-0025 TS44801-5000
Potassium hydrogen carbonate 99.5% for biochemistry
Potassium hydrogen carbonate
Formula: KHCO₃
Melting Pt: 292 °C
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Solubility Clear colorless
UV at 280 nm A: ≤0.05
Titration with HClO4 99.4 to 100.6 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.0005 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR