Sodium benzoate 99% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoic acid sodium salt

44780-0010 44780-2500
TS44780-0010EA 94 USD
TS44780-0010 TS44780-2500
Sodium benzoate 99% for biochemistry
Sodium benzoate
Formula: C₆H₅COONa
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline or granular powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Solubility (1 M in water) Clear colorless to light yellow
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤0.05 %
Sulfate (SO4) ≤0.05 %
Arsenic (As) ≤0.0002 %
Copper (Cu) ≤0.001 %
Mercury (Hg) ≤0.0001 %
Zinc (Zn) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR