Disodium hydrogen phosphate dodecahydrate 99% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate

44725-5000 44725-0025
TS44725-5000EA 130 USD
TS44725-5000 TS44725-0025
Disodium hydrogen phosphate dodecahydrate 99% for biochemistry
di-Sodium hydrogen phosphate dodecahydrate
Formula: Na₂HPO₄·12H₂O
MW: 358.14 g/mol
Melting Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS Number: 10039-32-4

Order Now

Specification Test Results

Appearance White Crystals
Solubility Clear colorless
UV (0.1 M in water)
pH 9.0 to 9.4 (0.1 M in water)
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤50 ppm
Arsenic (As) ≤0.5 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR