N-Methylisopropylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

43965-0100 43965-0500
TS43965-0100EA 77 USD
TS43965-0100 TS43965-0500
N-Methylisopropylamine 98%
N-Methylisopropylamine
Formula: C₄H₁₁N
Boiling Pt: 50…53 °C (760 mmHg)
Density: 0.702 g/cm³
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00042859
CAS Number: 4747-21-1
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20°C, 589 nm)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR