Ethylenediamine 99+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,2-Ethanediamine, 1,2-Diaminoethane
42726-1000
42726-8000
TS42726-1000EA
102
USD
TS42726-1000
TS42726-8000
Ethylenediamine 99+%, AcroSeal™
Ethylenediamine
Formula:
C₂H₈N₂ Boiling Pt: 117 °C (1013 hPa) Melting Pt: 11 °C Density: 0.898 g/cm³ Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008204 CAS Number: 107-15-3 UN: 1604 ADR: 8,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4555 to 1.4575 (20°C, 589 nm) |
Water | ≤0.5 % (Coulometric) |
GC | ≥99.0 % |
Color scale | ≤30 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...