Benzylamine 99.5+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Aminotoluene

42723-1000 42723-8000
TS42723-1000EA 92 USD
TS42723-1000 TS42723-8000
Benzylamine 99.5+%, AcroSeal™
Benzylamine
Formula: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Boiling Pt: 185
Melting Pt: 10
Density: 0.98 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
UN: 2735
ADR: 8 II
Merck Index: 13,01127

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5424 to 1.5444 (20°C, 589 nm)
Water ≤0.3 %
GC ≥99.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR