Sebaconitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

41925-0250 41925-1000
TS41925-0250EA 74 USD
TS41925-0250 TS41925-1000
Sebaconitrile 98%
Sebaconitrile
Formula: C₁₀H₁₆N₂
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.91 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00019900
CAS Number: 1871-96-1
EINECS: 217-494-0
UN: 3276

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4490 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR