Methyl-3-aminobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-aminobenzoate

41395-0050 41395-0250
TS41395-0050EA 79 USD
TS41395-0050 TS41395-0250
Methyl-3-aminobenzoate 98%
Methyl 3-aminobenzoate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 120
Melting Pt: 52…55 °C
MDL Number: MFCD00017102
CAS Number: 4518-10-9
EINECS: 224-842-5
UN: 2811

Order Now

Specification Test Results

Appearance Light beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 49 to 54 °C
Titration with HClO4 ≥97.5 %
Enantiomeric excess ≥97.5 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR