Isoamyl alcohol 99%, AcroSeal™ for biochemistry
Supplier: Thermo Scientific Chemicals
|
Warning
|
41272-0010
41272-0025
41272-1000
TS41272-0010EA
94
USD
TS41272-0010
TS41272-0025
TS41272-1000
Isoamyl alcohol 99%, AcroSeal™ for biochemistry
Isoamyl alcohol
Formula:
(CH₃)₂CHCH₂CH₂OH Boiling Pt: 130 °C (760 mmHg) Melting Pt: –70 °C Density: 0.8 g/cm³ Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 UN: 1105 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4050 to 1.4070 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥98.5 % |
UV | at 280 nm A: ≤0.1 |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...