2-Imidazolidinone, anhydrous 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Imidazolidone

41227-0010 41227-0250 41227-2500
TS41227-0010EA 331 USD
TS41227-0010 TS41227-0250 TS41227-2500
2-Imidazolidinone, anhydrous 96%
2-Imidazolidinone
Formula: C₃H₆N₂O
Boiling Pt: 187 °C (10Torr)
Melting Pt: 131.4…132.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005257
CAS Number: 120-93-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥95.0 %
GC ≥99.75 %
UV at 255 nm A: ≤1.0
Color scale ≤10 APHA
Specific gravity (20°C) 0.900 to 0.904
Residue after evaporation ≤0.0002 %
Acidity ≤0.0008 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR