Dextrin

Supplier: Thermo Scientific Chemicals

Synonyms: Dextrin

40628-1000 40628-5000
TS40628-1000EA 29 USD
TS40628-1000 TS40628-5000
Dextrin
Dextrin
Formula: (C₆H₁₂O₆)n
Storage Temperature: Ambient
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 13,02971

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.5 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR