Corn oil
Supplier: Thermo Scientific Chemicals
40543-0025
40543-5000
TS40543-0025EA
120
USD
TS40543-0025
TS40543-5000
Corn oil
Corn oil
Density:
0.92 g/cm³ Flash Pt: >200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130865 CAS Number: 8001-30-7 |
Specification Test Results
Appearance | Clear yellow Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4730 to 1.4760 (20°C, 589 nm) |
Acid value | ≤0.5 mg KOH/g |
Peroxide value | ≤5 meq O2/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...