2-Nitroethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

39796-0050 39796-0250 39796-1000
TS39796-0050EA 65 USD
TS39796-0050 TS39796-0250 TS39796-1000
2-Nitroethanol 97%
2-Nitroethanol
Formula: C₂H₅NO₃
MW: 91.07 g/mol
Boiling Pt: 194
Melting Pt: -80
Density: 1.292 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007405
CAS Number: 625-48-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR