N,N-Diethyl-p-phenylenediammonium sulfate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-N,N-diethylaniline sulfate, N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, N1,N1-Diethylbenzene-1,4-diamine sulfate, 4-Amino-N,N-diethylaniline sulphate, N,N-Diethyl-1,4-phenylenediammonium sulfate, N,N-Diethyl-p-phenylenediamine sulfate, N,N-Diethylbenzene-1,4-diammonium sulfate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate

39500-0010 39500-0500 39500-2500
TS39500-0010EA 312 USD
TS39500-0010 TS39500-0500 TS39500-2500
N,N-Diethyl-p-phenylenediammonium sulfate 99%
N,N-Diethyl-p-phenylenediammonium sulphate
Formula: C₁₀H₁₈N₂O₄S
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00012993
CAS Number: 6283-63-2
EINECS: 228-500-6

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals and/or lumps
Infrared spectrum Conforms
Melting point 184 to 187 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR