Diisopropylamine 99.5%, AcroSeal™, redistilled

Supplier: Thermo Scientific Chemicals
Danger

37830-0010 37830-1000
TS37830-0010EA 401 USD
TS37830-0010 TS37830-1000
Diisopropylamine 99.5%, AcroSeal™, redistilled
Diisopropylamine
Formula: C₆H₁₅N
Boiling Pt: 84 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.716 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008862
CAS Number: 108-18-9
EINECS: 203-558-5
UN: 1158
ADR: 3,II
Merck Index: 13,03219

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3930 (20°C, 589 nm)
GC ≥99.4 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR