Orotic acid, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Warning

35288-0100 35288-1000
TS35288-0100EA 43 USD
TS35288-0100 TS35288-1000
Orotic acid, anhydrous 98%
Orotic acid
Formula: C₅H₄N₂O₄
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006027
CAS Number: 65-86-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR