Triethylammonium dihydrogentrifluoride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Triethylamine trihydrofluoride, Triethylamine trishydrofluoride
35062-0250
35062-1000
TS35062-0250EA
87
USD
TS35062-0250
TS35062-1000
Triethylammonium dihydrogentrifluoride 97%
Triethylammonium dihydrogentrifluoride
Formula:
C₆H₁₈F₃N MW: 161.21 Boiling Pt: 70 °C (15 mmHg) Melting Pt: –29 °C Density: 0.989 g/cm³ Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043294 CAS Number: 73602-61-6 EINECS: 277-550-5 UN: 2922 ADR: 8 I |
Specification Test Results
Appearance | Colorless to yellow to orange Liquid |
Infrared spectrum | Conforms |
Titration with NaOH | 20 to 25 % (available HF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...