Triethylammonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triethylamine hydrobromide

34972-1000 34972-5000
TS34972-1000EA 41 USD
TS34972-1000 TS34972-5000
Triethylammonium bromide 99%
Triethylammonium bromide
Formula: C₆H₁₅N·HBr
MW: 182.1
Flash Pt: >65 °C
MDL Number: MFCD00054287
CAS Number: 636-70-4
EINECS: 211-263-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR